Sodium 4-aminohippurate hydrate 98%

A15775.06 A15775.14 A15775.22
A15775.06EA 37.1 EUR
A15775.06 A15775.14 A15775.22
Sodium 4-aminohippurate hydrate 98%
Sodium 4-aminohippurate hydrate

Purity: 98% CAS No.: 94-16-6 Molecular Formula: C9H9N2NaO3.xH2O Formula Weight: 216.17(anhy) Melting Point: 123-127° MDL No.: MFCD00150723

Formula: C₉H₉N₂NaO₃·H₂O
Melting Pt: 123…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00044689
CAS Number: 94-16-6
EINECS: 202-309-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR