2-Ethylbutyryl chloride 99%
Supplier: ACROS ORGANICS
|
Danger
|
409821000
409820050
409820250
ACRO409821000EA
248
EUR
ACRO409821000
ACRO409820050
ACRO409820250
2-Ethylbutyryl chloride 99%
2-Ethylbutyryl chloride
Formula:
C₆H₁₁ClO MW: 134.61 g/mol Flash Pt: 33 °C Storage Temperature: Ambient |
MDL Number:
MFCD00018812 CAS Number: 2736-40-5 EINECS: 220-359-9 UN: 2920 ADR: 8 II |
Specification Test Results
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4230 to 1.4250 (20°C, 589 nm) |
Titration RCOCl | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...