Methyl diphenylacetate ≥98%

Supplier: ALFA AESAR

B24694.06 B24694.14 B24694.22
B24694.06EA 50.7 EUR
B24694.06 B24694.14 B24694.22
Methyl diphenylacetate ≥98%
Methyl diphenylacetate

Purity: 98% CAS No.: 3469-00-9 Molecular Formula: (C6H5)2CHCO2CH3 Formula Weight: 226.28 Melting Point: 59-61° MDL No.: MFCD00025869

Formula: C₁₅H₁₄O₂
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00025869
CAS Number: 3469-00-9
EINECS: 222-431-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR