Hygiene Safety Ankle Boots, Slip-On
Supplier: RODO GROUP
SRC01-10 SRC01-11 SRC01-12 SRC01-13 SRC01-3 SRC01-5 SRC01-6 SRC01-7 SRC01-8 SRC01-9
114-4941EA 0 EUR
114-4941 114-4942 114-4943 114-4944 114-4945 114-4947 114-4948 114-4949 114-4950 114-4951
Hygiene Safety Ankle Boots, Slip-On
Shoes and Boots
Safety ankle boots made from water resistant microfibre upper. Self cleaning, lightweight PU sole (resistant to oils, fats, acids and alkalis, food and animal waste).
  • Anti-static, flexible sole with high slip resistance
  • Breathable and fast drying mesh lining
  • Padded ankle collar
  • Machine washable at 40 °C
  • Protective steel toecap (200 J)
Specially designed for cleanrooms, medical and food industries.
Colour: White.
Certifications: EN 20345; S2, SRC