MINO Digital Ovens
Supplier: GENLAB
MINO/30/F/DIG MINO/100/F/DIG MINO/125/F/DIG MINO/150/F/DIG MINO/200/F/DIG MINO/50/F/DIG MINO/75/F/DIG
MINO/30/F/DIGEA 1640 EUR
MINO/30/F/DIG 466-0436 466-0437 466-0438 466-0439 466-0441 466-0442
MINO Digital Ovens
Ovens Standard Ovens
Reliable, efficient, general purpose ovens, to suit most drying, warming and general laboratory applications. These units from Genlab have a sheet steel exterior finished with an easy to clean powder coated paint. The interior chamber is made from mild steel coated with aluminium (CLAD). They are heated via Incoloy sheathed elements fitted around the fan on the rear of the chamber. Ovens have fixed shelf runners and removable chrome plated wire grid shelves. The top vent is fitted with a clip to hold a glass immersion thermometer.
  • Temperature fluctuation ±0,5 °C
  • Temperature range 40
More Product Information