Anti-IL2RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BC96]

Supplier: Tonbo Biosciences
20-0259-T100 20-0259-T025
20-0259-T100EA 114 EUR
20-0259-T100 20-0259-T025
Anti-IL2RA Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: BC96]
Antibodies
The BC96 antibody is specific for human CD25, a 55 kDa surface protein also known as the Interleukin-2 Receptor alpha chain, or IL-2R alpha. CD25 may bind IL-2 by itself, although with low affinity and without induction of cell signaling. CD25 is also expressed within a high-affinity complex, along with the IL-2R beta chain (CD122) and the common gamma chain (CD132), to form a signaling receptor complex. Signaling via CD25 may modulate Th17 and Treg cell differentiation, and has been shown to play a role in the function of dendritic cells.

Type: Primary
Antigen: IL2RA (interleukin 2 receptor, alpha)
Clonality: monoclonal
Clone: BC96
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR