Anti-KLRB1 Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: PK136]

Supplier: Tonbo Biosciences
65-5941-U100 65-5941-U025
65-5941-U100EA 196 EUR
65-5941-U100 65-5941-U025
Anti-KLRB1 Mouse Monoclonal Antibody (Peridinin Chlorophyll/Cy5.5®) [clone: PK136]
Antibodies
The PK136 antibody is specific for mouse NK1.1, a type II transmembrane lectin-like receptor and member of the killer cell lectin-like receptor (KLR) family. NK1.1 is prominently expressed on natural killer (NK) cells, and is correlated with NK cytotoxic effects toward virus-infected cells and tumor cells. NK1.1 is expressed on subsets of NKT cells in certain mouse strains (C57BL/6, FVB/N, and NZB), yet absent from others (AKR, BALB/c, CBA/J, C3H, DBA/1, DBA/2, NOD, SJL, and 129). Putative subsets of NK cells and their expression of NK1.1 antigen are of continuing interest, including NK1.1+/CD117+ (c-Kit) cells reported to be immunosuppressive for CD8+ T cells in a mechanism involving PD-1 and PD-L1 (Ehlers et al. 2012. Endocrinology. 10: 1247).

The PK136 antibody may be used for detection of NK1.1 expression on mouse strains including CE, B6, NZB, C58, Ma/My, ST, SJL, and FVB. The antibody is reported to react with an epitope common to NKR-P1B and NKR-P1C alloantigenic forms of NK1.1 (Carlyle et al. 2006. J. Immunol. 176: 7511-7524).

Type: Primary
Antigen: KLRB1 (killer cell lectin-like receptor subfamily B, member 1)
Clonality: monoclonal
Clone: PK136
Conjugation: Peridinin Chlorophyll/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG2a Kappa
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR