Anti-TSEN2 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB1587-100 ORB1587-200
BIRBORB1587-100EA 0 EUR
BIRBORB1587-100 BIRBORB1587-200
Anti-TSEN2 Mouse Monoclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: TSEN2 (TSEN2 tRNA splicing endonuclease subunit)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR