Anti-ASMTL Rabbit polyclonal antibody

Supplier: Biorbyt
ORB1634-100 ORB1634-200
BIRBORB1634-100EA 0 EUR
BIRBORB1634-100 BIRBORB1634-200
Anti-ASMTL Rabbit polyclonal antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: ASMTL (acetylserotonin O-methyltransferase-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR