Anti-MID1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB1836-100 ORB1836-200
BIRBORB1836-100EA 0 EUR
BIRBORB1836-100 BIRBORB1836-200
Anti-MID1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: MID1 (midline 1 (Opitz/BBB syndrome))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR