Anti-SEMCAP3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB1922-100 ORB1922-200
BIRBORB1922-100EA 312 EUR
BIRBORB1922-100 BIRBORB1922-200
Anti-SEMCAP3 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: SEMCAP3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR