Anti-CPXCR1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2104-100 ORB2104-200
BIRBORB2104-100EA 0 EUR
BIRBORB2104-100 BIRBORB2104-200
Anti-CPXCR1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CPXCR1 (CPX chromosome region, candidate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR