Anti-G protein alpha Rabbit polyclonal antibody

Supplier: Biorbyt
ORB2176-100 ORB2176-200
BIRBORB2176-100EA 0 EUR
BIRBORB2176-100 BIRBORB2176-200
Anti-G protein alpha Rabbit polyclonal antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: G protein alpha
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR