Anti-KLHL11 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2236-100 ORB2236-200
BIRBORB2236-100EA 0 EUR
BIRBORB2236-100 BIRBORB2236-200
Anti-KLHL11 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: KLHL11 (kelch-like family member 11)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR