Anti-CCDC89 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB2488-100 ORB2488-200
BIRBORB2488-100EA 0 EUR
BIRBORB2488-100 BIRBORB2488-200
Anti-CCDC89 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:100-500; Immunochemistry: 1:50-250 (based on 0.5 mg/ml)

Type: Primary
Antigen: CCDC89 (coiled-coil domain containing 89)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR