Anti-CHRNB2 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB6468-100 ORB6468-200
BIRBORB6468-100EA 0 EUR
BIRBORB6468-100 BIRBORB6468-200
Anti-CHRNB2 Rabbit polyclonal antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CHRNB2 (cholinergic receptor, nicotinic, beta 2 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR