Anti-Melamine (1B12) Mouse Monoclonal Antibody [clone: 1B12]

Supplier: Biorbyt
ORB9025-100 ORB9025-200
BIRBORB9025-100EA 0 EUR
BIRBORB9025-100 BIRBORB9025-200
Anti-Melamine (1B12) Mouse Monoclonal Antibody [clone: 1B12]
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Melamine (1B12)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR