Anti-HIV gp41 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB10820-100 ORB10820-20 ORB10820-5 ORB10820-50
BIRBORB10820-100EA 0 EUR
BIRBORB10820-100 BIRBORB10820-20 BIRBORB10820-5 BIRBORB10820-50
Anti-HIV gp41 Rabbit polyclonal antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: HIV gp41
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR