Anti-CHRNA7 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB11096-100 ORB11096-200
BIRBORB11096-100EA 0 EUR
BIRBORB11096-100 BIRBORB11096-200
Anti-CHRNA7 Rabbit polyclonal antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CHRNA7 (cholinergic receptor, nicotinic, alpha 7 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR