Anti-VSIG4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB11565-100 ORB11565-200
BIRBORB11565-100EA 0 EUR
BIRBORB11565-100 BIRBORB11565-200
Anti-VSIG4 Rabbit Polyclonal Antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: VSIG4 (V-set and immunoglobulin domain containing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR