Anti-BSG Mouse Monoclonal Antibody (PE (Phycoerythrin))

Supplier: Biorbyt
ORB12140-100 ORB12140-50
BIRBORB12140-100EA 0 EUR
BIRBORB12140-100 BIRBORB12140-50
Anti-BSG Mouse Monoclonal Antibody (PE (Phycoerythrin))
Antibodies
Type: Primary
Antigen: BSG (basigin (Ok blood group))
Clonality: monoclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR