Anti-CHRNA2 Rabbit polyclonal antibody

Supplier: Biorbyt
ORB13597-100 ORB13597-20 ORB13597-200 ORB13597-5
BIRBORB13597-100EA 0 EUR
BIRBORB13597-100 BIRBORB13597-20 BIRBORB13597-200 BIRBORB13597-5
Anti-CHRNA2 Rabbit polyclonal antibody
Antibodies
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: CHRNA2 (cholinergic receptor, nicotinic, alpha 2 (neuronal))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR