Anti-CSK Goat Polyclonal Antibody

Supplier: Biorbyt
ORB18668-100 ORB18668-20
BIRBORB18668-100EA 0 EUR
BIRBORB18668-100 BIRBORB18668-20
Anti-CSK Goat Polyclonal Antibody
Antibodies
ELISA: 1:32000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: CSK (c-src tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR