Anti-CDCP1 Goat Polyclonal Antibody

Supplier: Biorbyt
ORB18721-100 ORB18721-20
BIRBORB18721-100EA 0 EUR
BIRBORB18721-100 BIRBORB18721-20
Anti-CDCP1 Goat Polyclonal Antibody
Antibodies
ELISA: 1:32000; Western Blot: 1-3 ?g/ml

Type: Primary
Antigen: CDCP1 (CUB domain containing protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR