Anti-SERPINE1 Goat Polyclonal Antibody

Supplier: Biorbyt
ORB18991-100 ORB18991-20
BIRBORB18991-100EA 0 EUR
BIRBORB18991-100 BIRBORB18991-20
Anti-SERPINE1 Goat Polyclonal Antibody
Antibodies
ELISA: 1:64000; Western Blot: 0.1-0.3 ?g/ml

Type: Primary
Antigen: SERPINE1 (serpin peptidase inhibitor, clade E (nexin, plasminogen activator inhibitor type 1), member 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR