Anti-PIN1 Mouse Monoclonal Antibody

Supplier: Biorbyt
ORB22779-100 ORB22779-20 ORB22779-5
BIRBORB22779-100EA 0 EUR
BIRBORB22779-100 BIRBORB22779-20 BIRBORB22779-5
Anti-PIN1 Mouse Monoclonal Antibody
Antibodies
Type: Primary
Antigen: PIN1 (peptidylprolyl cis/trans isomerase, NIMA-interacting 1)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 (Mouse)
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR