Anti-GRIA3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB49056-100 ORB49056-200
BIRBORB49056-100EA 0 EUR
BIRBORB49056-100 BIRBORB49056-200
Anti-GRIA3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: GRIA3 (glutamate receptor, ionotropic, AMPA 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG (Rabbit)
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR