Anti-BRAF Rabbit polyclonal antibody

Supplier: Biorbyt
ORB106374-100 ORB106374-20
BIRBORB106374-100EA 0 EUR
BIRBORB106374-100 BIRBORB106374-20
Anti-BRAF Rabbit polyclonal antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: BRAF (v-raf murine sarcoma viral oncogene homolog B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR