Anti-DOK1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB106524-100 ORB106524-20
BIRBORB106524-100EA 0 EUR
BIRBORB106524-100 BIRBORB106524-20
Anti-DOK1 Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000; Immunochemistry: 1:50-1:200

Type: Primary
Antigen: DOK1 (docking protein 1, 62kDa (downstream of tyrosine kinase 1))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR