Anti-VEGFB Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB106553-100 ORB106553-20
BIRBORB106553-100EA 0 EUR
BIRBORB106553-100 BIRBORB106553-20
Anti-VEGFB Rabbit Polyclonal Antibody
Antibodies
Western Blot: 1:500-1:2000

Type: Primary
Antigen: VEGFB (vascular endothelial growth factor B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR