Anti-RABL2A Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125336-100 ORB125336-200
BIRBORB125336-100EA 0 EUR
BIRBORB125336-100 BIRBORB125336-200
Anti-RABL2A Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RABL2A (RAB, member of RAS oncogene family-like 2A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR