Anti-ALDH1B1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125412-100 ORB125412-200
BIRBORB125412-100EA 0 EUR
BIRBORB125412-100 BIRBORB125412-200
Anti-ALDH1B1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ALDH1B1 (aldehyde dehydrogenase 1 family, member B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR