Anti-CCNB1IP1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125520-100 ORB125520-200
BIRBORB125520-100EA 0 EUR
BIRBORB125520-100 BIRBORB125520-200
Anti-CCNB1IP1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CCNB1IP1 (cyclin B1 interacting protein 1, E3 ubiquitin protein ligase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR