Anti-CPB2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125615-100 ORB125615-200
BIRBORB125615-100EA 0 EUR
BIRBORB125615-100 BIRBORB125615-200
Anti-CPB2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CPB2 (carboxypeptidase B2 (plasma))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR