Anti-FOXN4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125748-100 ORB125748-200
BIRBORB125748-100EA 0 EUR
BIRBORB125748-100 BIRBORB125748-200
Anti-FOXN4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FOXN4 (forkhead box N4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR