Anti-IL4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB125889-100 ORB125889-200
BIRBORB125889-100EA 0 EUR
BIRBORB125889-100 BIRBORB125889-200
Anti-IL4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: IL4 (interleukin 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR