Anti-RIN3 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126310-100 ORB126310-200
BIRBORB126310-100EA 0 EUR
BIRBORB126310-100 BIRBORB126310-200
Anti-RIN3 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RIN3 (Ras and Rab interactor 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR