Anti-RAB11FIP2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126655-100 ORB126655-200
BIRBORB126655-100EA 0 EUR
BIRBORB126655-100 BIRBORB126655-200
Anti-RAB11FIP2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RAB11FIP2 (RAB11 family interacting protein 2 (class I))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR