Anti-CTRO Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB126814-100 ORB126814-200
BIRBORB126814-100EA 0 EUR
BIRBORB126814-100 BIRBORB126814-200
Anti-CTRO Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: CTRO
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR