Anti-LIPB1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127155-100 ORB127155-200
BIRBORB127155-100EA 0 EUR
BIRBORB127155-100 BIRBORB127155-200
Anti-LIPB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: LIPB1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR