Anti-RSK1/2/3/4 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127205-100 ORB127205-200
BIRBORB127205-100EA 0 EUR
BIRBORB127205-100 BIRBORB127205-200
Anti-RSK1/2/3/4 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: RSK1/2/3/4
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR