Anti-ABL1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127360-100 ORB127360-200
BIRBORB127360-100EA 0 EUR
BIRBORB127360-100 BIRBORB127360-200
Anti-ABL1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ABL1 (c-abl oncogene 1, non-receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR