Anti-FYN Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127409-100 ORB127409-200
BIRBORB127409-100EA 0 EUR
BIRBORB127409-100 BIRBORB127409-200
Anti-FYN Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FYN (FYN oncogene related to SRC, FGR, YES)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR