Anti-ADARB1 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127640-100 ORB127640-200
BIRBORB127640-100EA 0 EUR
BIRBORB127640-100 BIRBORB127640-200
Anti-ADARB1 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: ADARB1 (adenosine deaminase, RNA-specific, B1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR