Anti-BHLHE41 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB127757-100 ORB127757-200
BIRBORB127757-100EA 0 EUR
BIRBORB127757-100 BIRBORB127757-200
Anti-BHLHE41 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: BHLHE41 (basic helix-loop-helix family, member e41)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR