Anti-EGFR Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128008-100 ORB128008-200
BIRBORB128008-100EA 0 EUR
BIRBORB128008-100 BIRBORB128008-200
Anti-EGFR Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: EGFR (epidermal growth factor receptor)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR