Anti-FGFR1/2 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128060-100 ORB128060-200
BIRBORB128060-100EA 0 EUR
BIRBORB128060-100 BIRBORB128060-200
Anti-FGFR1/2 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: FGFR1/2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR