Anti-HTR6 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128208-100 ORB128208-200
BIRBORB128208-100EA 0 EUR
BIRBORB128208-100 BIRBORB128208-200
Anti-HTR6 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: HTR6 (5-hydroxytryptamine (serotonin) receptor 6, G protein-coupled)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR