Anti-MAP3K7 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128319-100 ORB128319-200
BIRBORB128319-100EA 0 EUR
BIRBORB128319-100 BIRBORB128319-200
Anti-MAP3K7 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: MAP3K7 (mitogen-activated protein kinase kinase kinase 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR