Anti-PEX14 Rabbit Polyclonal Antibody

Supplier: Biorbyt
ORB128514-100 ORB128514-200
BIRBORB128514-100EA 0 EUR
BIRBORB128514-100 BIRBORB128514-200
Anti-PEX14 Rabbit Polyclonal Antibody
Antibodies
Type: Primary
Antigen: PEX14 (peroxisomal biogenesis factor 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR